Learn More
AVANTOR MASTERFLEX (VWR LLC) 316 SS fittings; female luer x 1/8"" hose barb
Catalog No. NC0783351
Encompass
Cadence 316 SS fittings; female luer x 1/8" hose barb, Exceptional durabilityEasy to disassemble for cleaningIdeal for harsh chemical environmentsAutoclavable
Catalog No. NC0783351 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4150728
Cadence 316 SS fittings; female luer x 1/8" hose barb, Exceptional durabilityEasy to disassemble for cleaningIdeal for harsh chemical environmentsAutoclavable