Learn More
Chemglass Life Sciences Chemglass 31 mm Rotary Bearing Rebuild Kit for Mechanical Seal Stir Bearings
SDP
Catalog No. 502733533
Encompass_Preferred
This 31 mm rotary bearing rebuild kit is a replacement repair kit designed to service Chemglass mechanical seal stir bearings. The kit supplies the internal bearing components required to refurbish a 31 mm rotary bearing assembly; tools and lubricant are sold separately.
  • Size: 31 mm
  • Includes bearing seal, ball bearings, and needle roller bearings
  • Used to repair 31 mm mechanical seal stir bearings
  • Does not include tools
  • Requires a small amount of specialty lubricant, sold separately
Catalog No. 50-273-3533 Supplier Chemglass Life Sciences Supplier No. CG2077M331
This 31 mm rotary bearing rebuild kit is a replacement repair kit designed to service Chemglass mechanical seal stir bearings. The kit supplies the internal bearing components required to refurbish a 31 mm rotary bearing assembly; tools and lubricant are sold separately.
  • Size: 31 mm
  • Includes bearing seal, ball bearings, and needle roller bearings
  • Used to repair 31 mm mechanical seal stir bearings
  • Does not include tools
  • Requires a small amount of specialty lubricant, sold separately