Learn More
Chemglass Life Sciences 31mm Rotary Bearing Rebuild Kit with Tools
SDP
Catalog No. 5000980618
Encompass_Preferred
Repair kits for Chemglass CG-2077-M series mechanical seal stir bearings. Kits are available with or without tools. Each bearing repair requires a small amount of Krytox grease CG-GPL-205-01 which is purchased separately.Kits with tools include:Adjustable Pin Spanner WrenchRetaining Ring PliersBearing SealBall bearingsNeedle Roller BearingsKits without tools include:Bearing SealBall bearingsNeedle Roller BearingsItems Needed for Rebuild:Appropriately Sized Rebuild KitSpanner WrenchesHydraulic or Arbor PressClean RagsMeans of Vacuum TestingKrytox Oil & GreaseFine Scotch-brite
Catalog No. 50-009-80618 Supplier Chemglass Life Sciences Supplier No. CG2077M231
Repair kits for Chemglass CG-2077-M series mechanical seal stir bearings. Kits are available with or without tools. Each bearing repair requires a small amount of Krytox grease CG-GPL-205-01 which is purchased separately.Kits with tools include:Adjustable Pin Spanner WrenchRetaining Ring PliersBearing SealBall bearingsNeedle Roller BearingsKits without tools include:Bearing SealBall bearingsNeedle Roller BearingsItems Needed for Rebuild:Appropriately Sized Rebuild KitSpanner WrenchesHydraulic or Arbor PressClean RagsMeans of Vacuum TestingKrytox Oil & GreaseFine Scotch-brite