Learn More
Ace Glass, Inc. PTFE banana stir blade, 31x146mm, 2L flask size, 10mm shafts
SDP
Catalog No. 50979667
Encompass_Preferred
31x146mm PTFE banana shaped stir blade for 10mm O.D. button type stir shafts. 3mm thick blades have a number of perforations, and are designed to closely fit a 2L round bottom flask.
Catalog No. 50-979-667 Supplier Ace Glass, Inc. Supplier No. 808709
31x146mm PTFE banana shaped stir blade for 10mm O.D. button type stir shafts. 3mm thick blades have a number of perforations, and are designed to closely fit a 2L round bottom flask.