Learn More
Medchemexpress LLC (32-carbonyl)-RMC-5552 | 2382768-55-8 | 95.0% | 1776.11 | C93H134N10O24 | 5 MG
SDP
Catalog No. 5000222101
Encompass_Preferred
(32-Carbonyl)-RMC-5552 is a small-molecule research inhibitor of the mechanistic target of rapamycin (mTOR) that potently inhibits phosphorylation of mTORC1 and mTORC2 substrates. It is intended for biochemical and cellular studies of PI3K/Akt/mTOR signaling and is supplied with analytical characterization for research use.
  • Potent dual inhibition of mTORC1 and mTORC2.
  • Useful in biochemical and cellular pharmacology studies.
  • Characterized molecular properties and CAS identifier for traceability.
  • Available as small-scale dry packs and as a DMSO solution for convenient dosing.
  • Includes analytical data to support quality control.
Catalog No. 50-002-22101 Supplier Medchemexpress LLC Supplier No. HY1349035MG
(32-Carbonyl)-RMC-5552 is a small-molecule research inhibitor of the mechanistic target of rapamycin (mTOR) that potently inhibits phosphorylation of mTORC1 and mTORC2 substrates. It is intended for biochemical and cellular studies of PI3K/Akt/mTOR signaling and is supplied with analytical characterization for research use.
  • Potent dual inhibition of mTORC1 and mTORC2.
  • Useful in biochemical and cellular pharmacology studies.
  • Characterized molecular properties and CAS identifier for traceability.
  • Available as small-scale dry packs and as a DMSO solution for convenient dosing.
  • Includes analytical data to support quality control.