Learn More
Medchemexpress LLC (32-carbonyl)-RMC-5552 | 2382768-55-8 | MFCD32708541 | 95.4% | C93H134N10O24 | 10MG
SDP
Catalog No. 5000260085
Encompass_Preferred
(32-Carbonyl)-RMC-5552 is a synthetic small-molecule inhibitor of mTOR signaling for research use. It exhibits potent activity against mTORC1 and shows effects on mTORC2 substrates, enabling study of mTOR-dependent cellular processes in biochemical and cell-based assays.
  • Potent inhibitor of mTORC1 and active against mTORC2 substrates.
  • High purity (95.4%).
  • Suitable for biochemical and cell-based assays.
  • Supplied as a solid, white to off-white powder.
  • Soluble in DMSO for stock preparation.
  • Intended for laboratory research use only.
Catalog No. 50-002-60085 Supplier Medchemexpress LLC Supplier No. HY13490310MG
(32-Carbonyl)-RMC-5552 is a synthetic small-molecule inhibitor of mTOR signaling for research use. It exhibits potent activity against mTORC1 and shows effects on mTORC2 substrates, enabling study of mTOR-dependent cellular processes in biochemical and cell-based assays.
  • Potent inhibitor of mTORC1 and active against mTORC2 substrates.
  • High purity (95.4%).
  • Suitable for biochemical and cell-based assays.
  • Supplied as a solid, white to off-white powder.
  • Soluble in DMSO for stock preparation.
  • Intended for laboratory research use only.