Learn More
Access Technologies Luer Stub 32 Gauge Blunt Needle Adaptors, 50 per Pack
Catalog No. NC0333614
Encompass
32 gauge blunt needle adaptors 50/pk. Straw - Outer Diameter Inches/MM 0.009/0.25; Inner Diameter Inches/MM 0.003/0.09.
Catalog No. NC0333614 Supplier Access Technologies Supplier No. LSA32
32 gauge blunt needle adaptors 50/pk. Straw - Outer Diameter Inches/MM 0.009/0.25; Inner Diameter Inches/MM 0.003/0.09.