Learn More
Beckman Coulter CytoFLEX SRT, Flow Cytometer
Supplier:  Beckman Coulter C69620
Encompass
The Beckman Coulter CytoFLEX SRT is a benchtop flow cytometer designed for high-throughput research applications. It utilizes a violet laser and 3 fluorescence detectors to enable up to 13-color analysis. Key features include a sample throughput of up to 1,000 samples per hour, 6-peak rainbow beads for daily QC, and a dedicated sterile sampling module for sorting into plates or tubes under sterile conditions. The CytoFLEX SRT provides researchers with a powerful and versatile flow cytometry platform for high-content multiparametric analysis and sterile cell sorting.
Catalog No. NC2332499