Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Goo Gone Adhesive Remover, Pro-Power, 32 oz, 1 ea

Catalog No. 501112167
Encompass

Goo Gone Pro-Power Adhesive Remover is a powerful cleaner designed to remove a wide range of stubborn substances. It effectively dissolves asphalt, glue, grease, grill build-up, gum, masking tape, oil, paint, tar, and varnish from various surfaces.

  • Formulated as an orange gel with a citrus odor
  • Contains petroleum distillates and D-Limonene as active ingredients
  • Container size: 32 oz bottle
  • Flash point: 85 °C (185 °F)

Catalog No. 50-111-2167 Supplier MSC Supplier No. 00924217
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goo Gone Pro-Power Adhesive Remover is a powerful cleaner designed to remove a wide range of stubborn substances. It effectively dissolves asphalt, glue, grease, grill build-up, gum, masking tape, oil, paint, tar, and varnish from various surfaces.

  • Formulated as an orange gel with a citrus odor
  • Contains petroleum distillates and D-Limonene as active ingredients
  • Container size: 32 oz bottle
  • Flash point: 85 °C (185 °F)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.