Learn More
Container & Packaging Supply 32 oz white HDPE plastic round tub
SDP
Catalog No. NC1624267
Encompass_Preferred
The T020 is a 32 oz white HDPE plastic round tub. Tubs have wide mouths, tapered sides, and a slightly smaller base. When empty, they're nestable. When fitted with their appropriate flexible lids, they're stackable. Tubs are often used for dairy products like yogurt, cheese, butter spreads, cream cheese, and other products like jams, jellies, deli salads, salsas, cookie dough, and more. High density polyethylene (HDPE) is marked with the resin identification code of 2. HDPE has a high tensile strength, good resistance to chemicals, good impact resistance, highly rigid, and highly scratch resistant.
Catalog No. NC1624267 Supplier Container & Packaging Supply Supplier No. T020
The T020 is a 32 oz white HDPE plastic round tub. Tubs have wide mouths, tapered sides, and a slightly smaller base. When empty, they're nestable. When fitted with their appropriate flexible lids, they're stackable. Tubs are often used for dairy products like yogurt, cheese, butter spreads, cream cheese, and other products like jams, jellies, deli salads, salsas, cookie dough, and more. High density polyethylene (HDPE) is marked with the resin identification code of 2. HDPE has a high tensile strength, good resistance to chemicals, good impact resistance, highly rigid, and highly scratch resistant.