Learn More
Electron Microscopy Sciences 32% Paraformaldehyde (formaldehyde) aqueous solution, (1) 100 mL bottle
SDP
Catalog No. 50980495
Encompass_Preferred
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. 100 ml bottles.
Catalog No. 50-980-495 Supplier Electron Microscopy Sciences Supplier No. 15714S
 Restricted Item
Add to Cart
Add to Cart
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. 100 ml bottles.