Learn More
Ace Glass, Inc. Rotameter tube, size 32, with stainless steel float: 120 to1200 mL/min air flow, 3.0 to 30mL/min water flow, with glass float: 48-475 mL/min air flow, 0.5-6.0 mL/min water flow
SDP
Catalog No. 50974813
Encompass
Catalog No. 50-974-813 Supplier Ace Glass, Inc. Supplier No. 748117