Learn More
Chemglass Life Sciences Silicone Sealing Ring, fits GL-32 Cap, fits tubing O.D. 11.0mm to 13.0mm
SDP
Catalog No. CG19704
Encompass_Preferred
Silicone Sealing Ring, fits GL-32 Cap, fits tubing O.D. 11.0mm to 13.0mm. Seal is made from silicone with a vulcanized PTFE washer having an aperture to fit the listed tubing size. For use with CG-195 cap.
Catalog No. CG19704 Supplier Chemglass Life Sciences Supplier No. CG19704
Silicone Sealing Ring, fits GL-32 Cap, fits tubing O.D. 11.0mm to 13.0mm. Seal is made from silicone with a vulcanized PTFE washer having an aperture to fit the listed tubing size. For use with CG-195 cap.