Learn More
Chemglass Life Sciences Vacuum Trap, 32mm Body OD x 250mm Body Length, 13mm OD Side and Inner Tubes
SDP
Supplier:  Chemglass Life Sciences CG451002
Encompass_Preferred
Vacuum Trap, 32mm Body OD x 250mm Body Length, 13mm OD Side and Inner Tubes. One piece trap used in the removal of undesired vapors in the vacuum system by condensation. 32mm Body OD x 250mm Body Length, 13mm OD Side and Inner Tubes.
Catalog No. CG451002