Learn More
MSC Value Collection 02-0320 320 Grit, Silicon Carbide Sanding Sheet 11" Long x 9" Wide, Extra Fine Grade, A Weighted Paper Backing
Catalog No. NC1897242
Encompass
Value Collection 02-0320 320 Grit, Silicon Carbide Sanding Sheet 11" Long x 9" Wide, Extra Fine Grade, A Weighted Paper Backing
Catalog No. NC1897242 Supplier MSC Supplier No. 05893201
Value Collection 02-0320 320 Grit, Silicon Carbide Sanding Sheet 11" Long x 9" Wide, Extra Fine Grade, A Weighted Paper Backing