Learn More
METTLER TOLEDO pH Sensor InPro 3253i SG/225
Supplier:  METTLER TOLEDO 52005378
Encompass
The InPro 3253i SG/225 is a pre-pressurized, liquid filled, low maintenance pH & ORP sensor, combined with a temperature sensor, for in-line measurement in demanding applications. The 225 mm version is optimal for medium installation lengths.
Catalog No. NC9019695