Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDS Empore™ 3293 Cesium RAD DISK, 47 mm
SDP

Catalog No. 13110008 Shop All CDS Analytical Products
Change view
Click to view available options
Quantity:
20/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-110-008 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-110-008 Supplier CDS Empore™ Supplier No. 98040500674EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selectively bonding Cs+ cations from sample solutions. Used for nucleid species detection. Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples.

Empore™ RAD Disks provide an efficient alternative to conventional radiochemical sample preparation methods that use wet chemistry or packed columns.

  • Empore RAD disks provide a sample prep solution for large volume aqueous samples
  • The disk format provides a large surface area for sorbent/sample contact
  • Fast flow rates and high throughput may be realized with use of an Empore RAD disk
  • RAD disks form convenient direct counting sources when used in applicable test methods
  • Available with a variety of packings in 47 mm size
  • Approved to detect Cesium-37
  • Total Cs content: not exceed 10 mg
  • Method detection limit: 5 to 10 pCi/L (200 to 400 mBq/L)

Specifications

Bed Weight 500 mg
Format Disk
For Use With (Equipment) Vacuum Manifolds or Automated SPE Equipment
Particle Size 25 to 80 μm
Type Extraction Disk
Quantity 20/Pk.

To reduce the risks associated with environmental contamination, handling radiochemical materials or false negative results: Read, understand and follow all recommendations in the product MSDS and Instructions for Use.

To reduce the risks associated with radiochemical materials: Always wear the proper PPE (Personal Protection Equipment) when handling used product.

To reduce the risks associated with false negative results for radiochemicals: Always use the Empore™ Cesium Rad Disks by the expiration date.

To reduce the risks associated with environmental contamination: Dispose of used disks according to standard radiochemical procedures and regulations. Unused disks may be disposed of in accordance with standard procedure for laboratory waste.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.