Learn More
Cole-Parmer Irreversible Descending Low-Temperature Indicators, 32°F (0°C), 10 Pk
Catalog No. NC9550338 Shop All Cole-Parmer Products
Encompass_Preferred
These labels offer a highly reliable and irreversible indication of exposure to low temperatures. Labels clearly indicate when the product to which it is attached to has been subjected to harsh temperatures, which could have detrimental effects on refrigerated pharmaceuticals, foods, chemicals, paints, and adhesives. Available in both ascending and descending style Accuracy is ±1C of the rated temperature. Shelf life: one year after date of purchase.
Catalog No. NC9550338 Supplier Cole-Parmer Supplier No. 9030960
These labels offer a highly reliable and irreversible indication of exposure to low temperatures. Labels clearly indicate when the product to which it is attached to has been subjected to harsh temperatures, which could have detrimental effects on refrigerated pharmaceuticals, foods, chemicals, paints, and adhesives. Available in both ascending and descending style Accuracy is ±1C of the rated temperature. Shelf life: one year after date of purchase.