Learn More
Accutome 32g, 0.5" Hypodermic Needle, 100/pk
Catalog No. NC1768913
Encompass
32 gauge, sterile TSK SteriJect hypodermic needle measuring 0.5" without safety. Stainless steel needle with beveled point. Green polypropylene hub with luer locking connection. Manufactured by TSK Laboratory. Comes in a box of 100.
Catalog No. NC1768913 Supplier Accutome Supplier No. AX16509
32 gauge, sterile TSK SteriJect hypodermic needle measuring 0.5" without safety. Stainless steel needle with beveled point. Green polypropylene hub with luer locking connection. Manufactured by TSK Laboratory. Comes in a box of 100.