Learn More
Braintree Scientific 32G Intrathecal Catheter System, 11.7cm, accepts 27g ndl
SDP
Catalog No. NC9986020
Encompass
CS-32 Intrathecal Catheter System 32ga Mouse Catheter System - This catheter system offers the smallest diameter catheter/stylet product available for murine procedures. Ultra-thin diameter catheters for applications in mice. This 32ga murine catheter is a key component of a withdrawal/ delivery system specifically developed for use in mice and other small research animals. A significant achievement over standard polyethylene-based products, its material and small size combine to produce a minimally reactive catheter, thereby reducing catheter-induced artifacts that could obscure experimental observations. Suitable for cardiovascular, intracranial, and other applications. Includes: 11.7cm 32ga (0.010" OD/0.005" ID) polyurethane catheter. Bonded connection assembly flared to accept a 27ga needle 0.003" diameter Teflon-coated stainless steel stylet to facilitate placement. Individually packaged **Sold for research purposes only and not for human use**
Catalog No. NC9986020 Supplier Braintree Scientific Supplier No. CS32
CS-32 Intrathecal Catheter System 32ga Mouse Catheter System - This catheter system offers the smallest diameter catheter/stylet product available for murine procedures. Ultra-thin diameter catheters for applications in mice. This 32ga murine catheter is a key component of a withdrawal/ delivery system specifically developed for use in mice and other small research animals. A significant achievement over standard polyethylene-based products, its material and small size combine to produce a minimally reactive catheter, thereby reducing catheter-induced artifacts that could obscure experimental observations. Suitable for cardiovascular, intracranial, and other applications. Includes: 11.7cm 32ga (0.010" OD/0.005" ID) polyurethane catheter. Bonded connection assembly flared to accept a 27ga needle 0.003" diameter Teflon-coated stainless steel stylet to facilitate placement. Individually packaged **Sold for research purposes only and not for human use**