Learn More
Ace Glass, Inc. 32 liter graduated cylindrical jar with side indents
SDP
Catalog No. 50972774
Encompass
32L graduated cylindrical jar with side indents for easier handling. Made of heavy wall glass. Also available with poly safety coating. Not for use with heat, pressure, or vacuum applications.
Catalog No. 50-972-774 Supplier Ace Glass, Inc. Supplier No. 623332
32L graduated cylindrical jar with side indents for easier handling. Made of heavy wall glass. Also available with poly safety coating. Not for use with heat, pressure, or vacuum applications.