Learn More
Bellco Glass, Inc. 32mm 2-Port Assembly (Schott) For Small 32mm ID
SDP
Catalog No. 501945790
Encompass_Preferred
2-Port Assembly 32mm is for use on small ID 32mm side arms for spinner flask vessels. Attaches to side arms with polypropylene screw cap and silicone liner. 316 l stainless steel construction. MADE IN THE USA Polypropylene screw cap White silicone gasket BOTH TUBES: .187"OD X .117" ID, SUPPLY WITH 32mm S/CAP & WHITE SILIONE LINER, NOTE: THIS PORT ASSY WILL FIT ANY 50ml SCHOTT BOTTLE OR ANY 32mm SCREW
Catalog No. 50-194-5790 Supplier Bellco Glass, Inc. Supplier No. 563752000
2-Port Assembly 32mm is for use on small ID 32mm side arms for spinner flask vessels. Attaches to side arms with polypropylene screw cap and silicone liner. 316 l stainless steel construction. MADE IN THE USA Polypropylene screw cap White silicone gasket BOTH TUBES: .187"OD X .117" ID, SUPPLY WITH 32mm S/CAP & WHITE SILIONE LINER, NOTE: THIS PORT ASSY WILL FIT ANY 50ml SCHOTT BOTTLE OR ANY 32mm SCREW