Learn More
Ace Glass, Inc. Acrylic safety shield and aluminum safety cap for Michel-Miller chromatography columns. Cap is 32mm ID and fits#7 threaded columns. The shield is 25cm long.
SDP
Catalog No. 5097257
Encompass_Preferred
Catalog No. 50-972-57 Supplier Ace Glass, Inc. Supplier No. 579832