Learn More
Ace Glass, Inc. 32mm I.D. x 30cm L Acrylic Safety Shield
SDP
Catalog No. 50978427
Encompass_Preferred
32mm I.D. x 30cm L Acrylic Safety Shield with aluminum cap, for use with #11 threaded Michel-Miller chromatography columns. Safety shield insures safe operation at pressures to 300 psi. Shield wall thickness: 3mm. The top thread of the column is hung in a polyethylene cap; the cap is then held by the plastic shield. The shield should be secured with a clamp.
Catalog No. 50-978-427 Supplier Ace Glass, Inc. Supplier No. 579841
32mm I.D. x 30cm L Acrylic Safety Shield with aluminum cap, for use with #11 threaded Michel-Miller chromatography columns. Safety shield insures safe operation at pressures to 300 psi. Shield wall thickness: 3mm. The top thread of the column is hung in a polyethylene cap; the cap is then held by the plastic shield. The shield should be secured with a clamp.