Learn More
Premier Lab Supply Tapered Wall Aluminum Cups Producing 32mm Pellets – PN# AC-532
SDP
Catalog No. NC1976819
Encompass_Preferred
Tapered Wall Aluminum Cups with Dimensions 31mm x 7.9mm (PN# AC-532) – Suitable for producing 32mm pellets – PREMIER’s aluminum cups are made with a strong and durable aluminum alloy that provides stable backbone for pressed pellets. Thin wall material ensures minimal back pressure and eases reproducibility of pressed pellets. – 1,000 per bag.
Catalog No. NC1976819 Supplier Premier Lab Supply Supplier No. AC532
Tapered Wall Aluminum Cups with Dimensions 31mm x 7.9mm (PN# AC-532) – Suitable for producing 32mm pellets – PREMIER’s aluminum cups are made with a strong and durable aluminum alloy that provides stable backbone for pressed pellets. Thin wall material ensures minimal back pressure and eases reproducibility of pressed pellets. – 1,000 per bag.