Learn More
Chemglass Life Sciences 8" Upper Agitator Assembly 30mm Shaft, Improved
SDP
Catalog No. NC1400863
Encompass_Preferred
8" Upper Agitator Assembly 30mm Shaft, Improved. Agitators are made entirely of PTFE and glass filled PTFE and are for use with 30mm OD stirrer shaft CG-2097 in 75 and 100L reactors. The 45¿ angle impellers are set up to stir downwards in a clockwise direction and upwards when rotated counter-clockwise. The upper agitator hub is open so that it can be positioned wherever the indents are along the 30mm shaft. Locks onto shaft with a collet hub and nut and has a glass filled PTFE safety screw. Heavy-duty blades are 6.5mm thick.
Catalog No. NC1400863 Supplier Chemglass Life Sciences Supplier No. CG2095R51
8" Upper Agitator Assembly 30mm Shaft, Improved. Agitators are made entirely of PTFE and glass filled PTFE and are for use with 30mm OD stirrer shaft CG-2097 in 75 and 100L reactors. The 45¿ angle impellers are set up to stir downwards in a clockwise direction and upwards when rotated counter-clockwise. The upper agitator hub is open so that it can be positioned wherever the indents are along the 30mm shaft. Locks onto shaft with a collet hub and nut and has a glass filled PTFE safety screw. Heavy-duty blades are 6.5mm thick.