Learn More
Ace Glass, Inc. 32mm I.D. x 30cm L Acrylic Safety Shield, Quartz length is 7.6cm, and borosilicate length is 10.2cm.
SDP
Catalog No. 50974604
Encompass
32mm I.D. x 30cm L Acrylic Safety Shield for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-604 Supplier Ace Glass, Inc. Supplier No. 845542
32mm I.D. x 30cm L Acrylic Safety Shield for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.