Learn More
Chemglass Life Sciences Clamp, Horseshoe Style, 32mm Flange
SDP
Supplier:  Chemglass Life Sciences CG13901
Encompass_Preferred
Clamp, Horseshoe Style, 32mm Flange. Horseshoe shaped clamp used to connect CG-138 flat o-ring flange. Design allows clamp to be installed or removed easily. Made of cast aluminum with chemically resistant green epoxy coating. Three adjustable tension locking screws assure even clamping pressure.
Catalog No. CG13901