Learn More
Premier Lab Supply CEMBLES Pre-Assembled XRF Sample Cups – PN# PA-160-32-240
SDP
Catalog No. NC0282430
Encompass_Preferred
CEMBLES Pre-Assembled XRF Sample Cups! (CEMBLES PN# PA-160-32-240; 32mm Cups PN# SC-3332; Polypropylene film 4.0μm; 48 per pack). CEMBLES are custom made to your specifications & available in variant configurations of cups and thin films that have been pre-assembled for your convenience.
Catalog No. NC0282430 Supplier Premier Lab Supply Supplier No. PA16032240
CEMBLES Pre-Assembled XRF Sample Cups! (CEMBLES PN# PA-160-32-240; 32mm Cups PN# SC-3332; Polypropylene film 4.0μm; 48 per pack). CEMBLES are custom made to your specifications & available in variant configurations of cups and thin films that have been pre-assembled for your convenience.