Learn More
Sigma Aldrich Fine Chemicals Biosciences 32mm Round cell window KBr1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z1121781EA
Encompass
Cut and polished for demountable liquid and gas cell holders
Catalog No. 50-290-0333