Learn More
Cayman Chemical 3-[2-N-(Biotinyl)aminoethyldithio]propanoic Acid, Purity: ≥98%, CAS: 104582-29-8, 100 mg
SDP
Catalog No. 502970514
Encompass_Preferred
Features a biotin group and propanoic acid separated by a disulfide linkage; allows biotinylation of targets through either a chemical or enzymatic acylation reaction; disulfide bond permits removal of the biotin group with reducing agents
Catalog No. 50-297-0514 Supplier Cayman Chemical Supplier No. 16461100 MG
Features a biotin group and propanoic acid separated by a disulfide linkage; allows biotinylation of targets through either a chemical or enzymatic acylation reaction; disulfide bond permits removal of the biotin group with reducing agents