Learn More
C&G Containers 32OZ NAT W/M 89-400 L3
SDP
Catalog No. NC9516991
Encompass_Preferred
32oz Natural Wide/Mouth HDPE 89-400 white PP cap F217-lined assembled only (bottle with cap attached) L3 160/cs
Catalog No. NC9516991 Supplier C&G Containers Supplier No. LPW0329009
32oz Natural Wide/Mouth HDPE 89-400 white PP cap F217-lined assembled only (bottle with cap attached) L3 160/cs