Learn More
Cayman Chemical 33-BCRP Inhibitor, Purity: ≥98%, CAS: 2953730-35-1, 10 mg
SDP
Catalog No. 502916996
Encompass_Preferred
A BCRP inhibitor; sensitizes mitoxantrone-resistant H460/MX20 cells to mitoxantrone-induced cell death alone and in combination with UV radiation; sensitizes KB-C2 cells overexpressing P-gp to colchicine-induced cell death; increases intracellular accumulation of mitoxantrone in H460/MX20 cells at 5 µM
Catalog No. 50-291-6996 Supplier Cayman Chemical Supplier No. 3940310 MG
A BCRP inhibitor; sensitizes mitoxantrone-resistant H460/MX20 cells to mitoxantrone-induced cell death alone and in combination with UV radiation; sensitizes KB-C2 cells overexpressing P-gp to colchicine-induced cell death; increases intracellular accumulation of mitoxantrone in H460/MX20 cells at 5 µM