Learn More
Cayman Chemical 33-cGAMPsodIum salt 100ug
SDP
Catalog No. 502966981
Encompass_Preferred
A second messenger produced by bacteria that regulates chemotaxis, colonization, and other bacterial functions; binds weakly to STING (Kd = 1.04 µM); induces IFN-β mRNA expression in L929 cells (EC50 = 10.5 nM)
Catalog No. 50-296-6981 Supplier Cayman Chemical Supplier No. 17966100 UG
A second messenger produced by bacteria that regulates chemotaxis, colonization, and other bacterial functions; binds weakly to STING (Kd = 1.04 µM); induces IFN-β mRNA expression in L929 cells (EC50 = 10.5 nM)