Learn More
Ace Glass, Inc. Rotameter tube, size 33, with stainless steel float: 475 to5700 mL/min air flow, 10 to 180mL/min water flow, with glass float: 240-2800 mL/min air flow, 5-60 mL/min water flow
SDP
Catalog No. 50979259
Encompass
Catalog No. 50-979-259 Supplier Ace Glass, Inc. Supplier No. 748119