Learn More
Ace Glass, Inc. 330Mm Por B Tubes (Gb)
SDP
Catalog No. 509557729
Encompass_Preferred
330mm porosity B gas dispersion tube for use with #7 Ace-Threds. Tube is bent 15° to allow positioning inside flask to avoid stirring paddles, and reach close to the bottom of the flask.
Catalog No. 50-955-7729 Supplier Ace Glass, Inc. Supplier No. 720408
330mm porosity B gas dispersion tube for use with #7 Ace-Threds. Tube is bent 15° to allow positioning inside flask to avoid stirring paddles, and reach close to the bottom of the flask.