Learn More
Caron Products And Services CO2 Incubator, Reach-In, 33 cu. ft., Temp & RH (water pan)
Catalog No. NC1751025
Encompass_Preferred
Carons 934L (33 ft3) Reach-In CO2 Incubator. Standard features: Intuitive touchscreen controls, Precise IR CO2 Sensor, Controlled Temperature, Elevated Humidity and Tool-less Removable Interior. Multiple options available, including Automatic Decon Cycle and gVapor Controlled Humidity. 115V, 60Hz.
Catalog No. NC1751025 Supplier Caron Products And Services Supplier No. 7400331
Add to Cart
Add to Cart
Request a Quote
Carons 934L (33 ft3) Reach-In CO2 Incubator. Standard features: Intuitive touchscreen controls, Precise IR CO2 Sensor, Controlled Temperature, Elevated Humidity and Tool-less Removable Interior. Multiple options available, including Automatic Decon Cycle and gVapor Controlled Humidity. 115V, 60Hz.