Learn More
Caron Products And Services Heated Incubator. 33 cubic foot,
Catalog No. NC2308871
Encompass_Preferred
Heated Incubator. 33 cubic foot with temperature control, touchscreen. 10C above ambient to 70C, 115V, 60 Hz.
Catalog No. NC2308871 Supplier Caron Products And Services Supplier No. 7002331
Add to Cart
Add to Cart
Request a Quote
Heated Incubator. 33 cubic foot with temperature control, touchscreen. 10C above ambient to 70C, 115V, 60 Hz.