Learn More
Hunter Lab 33 mm Transmission Cell with Screw Top
SDP
Catalog No. NC1923969
Encompass
The 33-mm has a large easy-pour 22-mm OD/18-mm ID with a Teflon-lined, hard polypropylene screw-thread top.
Catalog No. NC1923969 Supplier Hunter Lab Supplier No. D121011891
The 33-mm has a large easy-pour 22-mm OD/18-mm ID with a Teflon-lined, hard polypropylene screw-thread top.