Learn More
American Radiolabeled Chemicals Inc 33P AS ORTHOPHOS ACID 1MCI
SDP
Catalog No. NC1714398
Encompass
PHOSPHORUS-33 AS ORTHOPHOSPHORIC ACID(H3PO4); 1mCi; Specific Activity: 3000 Ci/mmol 111 TBq/mmol; AMERICAN RADIOLABELED CHEMICALS INC mfg# ARP-0133-1
Catalog No. NC1714398 Supplier American Radiolabeled Chemicals Inc Supplier No. ARP01331
Add to Cart
Add to Cart
Request a Quote
PHOSPHORUS-33 AS ORTHOPHOSPHORIC ACID(H3PO4); 1mCi; Specific Activity: 3000 Ci/mmol 111 TBq/mmol; AMERICAN RADIOLABELED CHEMICALS INC mfg# ARP-0133-1