Learn More
Ace Glass, Inc. Connecting adapter, enlarging, 34/45 outer top, 24/40 inner bottom
SDP
Catalog No. 50971596
Encompass_Preferred
Connecting, enlarging adapter featuring a 34/45 outer top female joint and a 24/40 inner bottom male joint.
Catalog No. 50-971-596 Supplier Ace Glass, Inc. Supplier No. 500528
Connecting, enlarging adapter featuring a 34/45 outer top female joint and a 24/40 inner bottom male joint.