Learn More
Chemglass Life Sciences 34/45 Inner Joint, Cut Drip, on 32mm OD x 2.4mm W tubing
SDP
Supplier:  Chemglass Life Sciences CG11718
Encompass_Preferred
34/45 Inner Joint, Cut Drip, on 32mm OD x 2.4mm W tubing. Drip tip. Joints are available uncut or with the drip tip being cut at a 45º angle. Tooled on the same tubing sizes as the CG-100 inner joints.
Catalog No. CG11718