Learn More
Ace Glass, Inc. 34/45 INNER JOINT (GB)
SDP
Catalog No. 50979335
Encompass_Preferred
34/45 standard taper inner member ground joint for use with ACE 7565, 7566, 7567, 7651 and 7652 outer members. May also be used with ACE 7568 and 7585 half joints.
Catalog No. 50-979-335 Supplier Ace Glass, Inc. Supplier No. 756542
34/45 standard taper inner member ground joint for use with ACE 7565, 7566, 7567, 7651 and 7652 outer members. May also be used with ACE 7568 and 7585 half joints.