Learn More
Chemglass Life Sciences Sleeve, PTFE, Full Length, 34/45 Joint Size
SDP
Catalog No. CG11617
Encompass_Preferred
Sleeve, PTFE, Medium Length, 34/45 Joint Size. Reusable .005in thick PTFE sleeve for standard taper joints provides a grease-free seal.
Catalog No. CG11617 Supplier Chemglass Life Sciences Supplier No. CG11617
Sleeve, PTFE, Medium Length, 34/45 Joint Size. Reusable .005in thick PTFE sleeve for standard taper joints provides a grease-free seal.