Learn More
Ace Glass, Inc. 34/45 PTFE Stopper with Polypropylene Extraction Nut, Solid
SDP
Catalog No. 50977610
Encompass
34/45 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.
Catalog No. 50-977-610 Supplier Ace Glass, Inc. Supplier No. 1263622
34/45 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.