Learn More
Chemglass Life Sciences Sleeve, PTFE, Ribbed, w/Gripping Ring, 34/45 Joint Size
SDP
Catalog No. 50872448
Encompass_Preferred
Sleeve, PTFE, Ribbed, w/Gripping Ring, 34/45 Joint Size. Reusable PTFE sleeve with knurled, reinforced gripping ring for easy removal. Sleeve has reinforcement ribs on the outer surface that provide a vacuum tight seal to 1 mbar and also prevent the sleeve from sticking. Sleeve is .008in thick.
Catalog No. 50-872-448 Supplier Chemglass Life Sciences Supplier No. CG13605
Sleeve, PTFE, Ribbed, w/Gripping Ring, 34/45 Joint Size. Reusable PTFE sleeve with knurled, reinforced gripping ring for easy removal. Sleeve has reinforcement ribs on the outer surface that provide a vacuum tight seal to 1 mbar and also prevent the sleeve from sticking. Sleeve is .008in thick.