Learn More
Organomation Unheated 34 Position N-EVAP Nitrogen Evaporator
SDP
Catalog No. NC1963650
Encompass_Preferred
Our unheated 34-position nitrogen evaporator (cat# 11634-O) offers a large number of sample positions, without compromising any of the classic N-EVAP features. This time-tested, durable system provides one of the least expensive prices per position of all our N-EVAPs. The evaporative system, along with the standard flow control enables the operator to achieve the desired level of concentration with efficient nitrogen usage. Our unique dual band spring hoist assembly ensures easy and effective operator use.
Catalog No. NC1963650 Supplier Organomation Supplier No. 11634O
Add to Cart
Add to Cart
Request a Quote
Our unheated 34-position nitrogen evaporator (cat# 11634-O) offers a large number of sample positions, without compromising any of the classic N-EVAP features. This time-tested, durable system provides one of the least expensive prices per position of all our N-EVAPs. The evaporative system, along with the standard flow control enables the operator to achieve the desired level of concentration with efficient nitrogen usage. Our unique dual band spring hoist assembly ensures easy and effective operator use.