Learn More
Beckman Coulter Beckman Coulter MET ONE 3423+ Portable Air Particle Counter, 50 L/min
Supplier:  Beckman Coulter C38681
Encompass
The MET ONE 3423+ portable air particle counter is designed for routine environmental monitoring in GMP cleanrooms. It delivers reliable particle counting across common cleanroom size channels, supports electronic SOP maps and guided sampling, and offers multiple connectivity options and battery operation for portable use.
  • 50 L per minute flow rate for rapid sampling
  • Detects particles in the 0.3-10 μm range
  • Simplifies SOP sampling with electronic maps and guided tracking
  • Built-in USB, wired Ethernet, and Wi-Fi for data transfer
  • Battery operation for up to 5.7 hours typical use
  • Compact, portable design with 5.6 kg weight (without battery)
Catalog No. NC2818085
Request a Quote
To request a quote for this item, please select Request a Quote to reach our dedicated support team.