Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Tube Thinwall Ultra-Clear 5mL 13x51mm (50pkg)

Catalog No. NC9157569
Encompass_Preferred

Ultra-Clear Tube, Open-Top,Thinwall, 5 mL, 13 x 51mm, Quantity: 50Pk

Catalog No. NC9157569 Supplier Beckman Coulter Supplier No. 344057
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ultra-Clear Tube, Open-Top,Thinwall, 5 mL, 13 x 51mm, Quantity: 50Pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.