Learn More
eMolecules​ 344591-91-9 | (1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl)boronic acid | Synthonix - Stock | MFCD08701785 | 193.920 | C5H6BF3N2O2 | 97.000 | Cn1nc(cc1B(O)O)C(F)(F)F | 1g | 459648377
Catalog No. 5000159594 Shop All eMolecules Inc. Products
Encompass_Preferred
(1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl)boronic acid | Synthonix - Stock | 344591-91-9 | MFCD08701785 | 193.920 | C5H6BF3N2O2 | 97.000 | Cn1nc(cc1B(O)O)C(F)(F)F | 1g | 459648377
Catalog No. 50-001-59594 Supplier eMolecules​ Supplier No. 459648377
(1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl)boronic acid | Synthonix - Stock | 344591-91-9 | MFCD08701785 | 193.920 | C5H6BF3N2O2 | 97.000 | Cn1nc(cc1B(O)O)C(F)(F)F | 1g | 459648377